Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
27 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM206 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TMEM206 - transmembrane protein 206 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198862.1 | Intron | NP_001185791.1 | ||||
NM_018252.2 | Intron | NP_060722.2 | ||||
XM_005273177.3 | Intron | XP_005273234.1 | ||||
XM_011509717.2 | Intron | XP_011508019.1 | ||||
XM_011509718.2 | Intron | XP_011508020.1 | ||||
XM_017001634.1 | Intron | XP_016857123.1 | ||||
XM_017001635.1 | Intron | XP_016857124.1 | ||||
XM_017001636.1 | Intron | XP_016857125.1 | ||||
XM_017001637.1 | Intron | XP_016857126.1 |