Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613367 MIM: 612537 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIN54 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LIN54 - lin-54 DREAM MuvB core complex component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001115007.2 | 2465 | UTR 3 | NP_001108479.1 | |||
NM_001115008.2 | 2465 | UTR 3 | NP_001108480.1 | |||
NM_001288996.1 | 2465 | UTR 3 | NP_001275925.1 | |||
NM_001288997.1 | 2465 | UTR 3 | NP_001275926.1 | |||
NM_194282.3 | 2465 | UTR 3 | NP_919258.2 | |||
XM_005262750.4 | 2465 | UTR 3 | XP_005262807.1 | |||
XM_006714081.3 | 2465 | UTR 3 | XP_006714144.1 | |||
XM_017007728.1 | 2465 | UTR 3 | XP_016863217.1 | |||
XM_017007729.1 | 2465 | UTR 3 | XP_016863218.1 | |||
XM_017007730.1 | 2465 | UTR 3 | XP_016863219.1 |
THAP9 - THAP domain containing 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |