Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTGCTCGGCCCCTGCTGGTCGCG[C/G]AGCATCTTCTCCAGCACGCCGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608137 MIM: 612122 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NSMF PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NSMF - NMDA receptor synaptonuclear signaling and neuronal migration factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PNPLA7 - patatin like phospholipase domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098537.2 | 4089 | Silent Mutation | CTC,CTG | L,L 1239 | NP_001092007.2 | |
NM_152286.4 | 4089 | Silent Mutation | CTC,CTG | L,L 1214 | NP_689499.4 | |
XM_006717102.1 | 4089 | Silent Mutation | CTC,CTG | L,L 1240 | XP_006717165.1 | |
XM_006717104.3 | 4089 | Silent Mutation | CTC,CTG | L,L 1206 | XP_006717167.1 | |
XM_011518664.2 | 4089 | Intron | XP_011516966.1 | |||
XM_017014709.1 | 4089 | Silent Mutation | CTC,CTG | L,L 1206 | XP_016870198.1 |