Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGAGCTTTTCCCCATAGAGAGTA[C/G]GTTGGAGATGTAGGGAAGCAAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600549 MIM: 602137 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IK - IK cytokine, down-regulator of HLA II | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006083.3 | Intron | NP_006074.2 |
MIR3655 - microRNA 3655 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFA2 - NADH:ubiquinone oxidoreductase subunit A2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMCO6 - transmembrane and coiled-coil domains 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |