Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTGGATTGGCTGGTGGGGAATTG[G/T]GGAACATCAAAAATAACTCTGCAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606627 MIM: 603707 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DAAM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DAAM2 - dishevelled associated activator of morphogenesis 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100505635 - uncharacterized LOC100505635 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MOCS1 - molybdenum cofactor synthesis 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001075098.3 | 1166 | UTR 3 | NP_001068566.1 | |||
NM_005943.5 | 1166 | UTR 3 | NP_005934.2 | |||
XM_011514632.2 | 1166 | Missense Mutation | CAC,CCC | H,P 390 | XP_011512934.1 | |
XM_011514633.2 | 1166 | Missense Mutation | CAC,CCC | H,P 374 | XP_011512935.1 | |
XM_011514634.2 | 1166 | Missense Mutation | CAC,CCC | H,P 303 | XP_011512936.1 | |
XM_017010888.1 | 1166 | UTR 3 | XP_016866377.1 |