Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608937 MIM: 602389 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH2B1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SH2B1 - SH2B adaptor protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145795.1 | 45 | UTR 5 | NP_001139267.1 | |||
NM_001145796.1 | 45 | UTR 5 | NP_001139268.1 | |||
NM_001145797.1 | 45 | UTR 5 | NP_001139269.1 | |||
NM_001145812.1 | 45 | Intron | NP_001139284.1 | |||
NM_001308293.1 | 45 | Intron | NP_001295222.1 | |||
NM_001308294.1 | 45 | UTR 5 | NP_001295223.1 | |||
NM_015503.2 | 45 | Intron | NP_056318.2 | |||
XM_017023114.1 | 45 | Intron | XP_016878603.1 | |||
XM_017023115.1 | 45 | Intron | XP_016878604.1 | |||
XM_017023116.1 | 45 | Intron | XP_016878605.1 | |||
XM_017023117.1 | 45 | Intron | XP_016878606.1 | |||
XM_017023118.1 | 45 | Intron | XP_016878607.1 | |||
XM_017023119.1 | 45 | UTR 5 | XP_016878608.1 | |||
XM_017023120.1 | 45 | Intron | XP_016878609.1 |
TUFM - Tu translation elongation factor, mitochondrial | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |