Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCACTGATGAAACCATTGGTCTCG[C/T]TGGCAGAGAGACTGCCAAAGTCAGC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIER3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIER3 - MIER family member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297598.1 | 1652 | Missense Mutation | AAC,AGC | N,S 535 | NP_001284527.1 | |
NM_001297599.1 | 1652 | Missense Mutation | AAC,AGC | N,S 530 | NP_001284528.1 | |
NM_152622.4 | 1652 | Missense Mutation | AAC,AGC | N,S 529 | NP_689835.3 | |
XM_011543216.1 | 1652 | Missense Mutation | AAC,AGC | N,S 546 | XP_011541518.1 | |
XM_011543217.1 | 1652 | Missense Mutation | AAC,AGC | N,S 545 | XP_011541519.1 | |
XM_017009149.1 | 1652 | Missense Mutation | AAC,AGC | N,S 354 | XP_016864638.1 |
SETD9 - SET domain containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171990.2 | 1652 | Intron | NP_001165461.1 | |||
NM_001323018.1 | 1652 | Intron | NP_001309947.1 | |||
NM_001323019.1 | 1652 | Intron | NP_001309948.1 | |||
NM_001323020.1 | 1652 | Intron | NP_001309949.1 | |||
NM_001323022.1 | 1652 | Intron | NP_001309951.1 | |||
NM_153706.3 | 1652 | Intron | NP_714917.2 | |||
XM_005248426.4 | 1652 | Intron | XP_005248483.1 | |||
XM_011543139.2 | 1652 | Intron | XP_011541441.1 | |||
XM_011543140.2 | 1652 | Intron | XP_011541442.1 |
Set Membership: |
HapMap |