Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCTGCAGCCCCAGTAGGCGTTGC[C/G]GGCACAGGAGCCTCAGCTGCCTCGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 300363 | ||||||||||||||||||||
Literature Links: |
ARMCX2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARMCX2 - armadillo repeat containing, X-linked 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282231.1 | 1221 | Silent Mutation | CCC,CCG | P,P 223 | NP_001269160.1 | |
NM_014782.6 | 1221 | Silent Mutation | CCC,CCG | P,P 223 | NP_055597.1 | |
NM_177949.3 | 1221 | Silent Mutation | CCC,CCG | P,P 223 | NP_808818.1 |