Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAGAAAGAGGAGAGATTTCCTAG[C/T]TCTGTAAGAACACCTAGTATTTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607489 MIM: 600589 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CUL9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CUL9 - cullin 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015089.3 | Intron | NP_055904.1 | ||||
XM_011514422.1 | Intron | XP_011512724.1 | ||||
XM_011514423.2 | Intron | XP_011512725.1 | ||||
XM_011514424.1 | Intron | XP_011512726.1 | ||||
XM_011514425.1 | Intron | XP_011512727.1 | ||||
XM_011514426.1 | Intron | XP_011512728.1 | ||||
XM_011514427.1 | Intron | XP_011512729.1 | ||||
XM_011514428.1 | Intron | XP_011512730.1 | ||||
XM_011514429.1 | Intron | XP_011512731.1 | ||||
XM_011514430.1 | Intron | XP_011512732.1 | ||||
XM_017010589.1 | Intron | XP_016866078.1 | ||||
XM_017010590.1 | Intron | XP_016866079.1 |
SRF - serum response factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |