Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATGATCTCTTCTGTAATTTGTGCA[A/G]TGCCACTCCTTTACATAATCTTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600227 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABCA17P PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ABCA17P - ATP binding cassette subfamily A member 17, pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CCNF - cyclin F | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323538.1 | Intron | NP_001310467.1 | ||||
NM_001761.2 | Intron | NP_001752.2 | ||||
XM_017023822.1 | Intron | XP_016879311.1 | ||||
XM_017023823.1 | Intron | XP_016879312.1 |
Set Membership: |
HapMap |