Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602354 MIM: 612583 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LAT - linker for activation of T-cells | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPNS1 - sphingolipid transporter 1 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142448.1 | Intron | NP_001135920.1 | ||||
NM_001142449.1 | Intron | NP_001135921.1 | ||||
NM_001142450.1 | Intron | NP_001135922.1 | ||||
NM_001142451.1 | Intron | NP_001135923.1 | ||||
NM_032038.2 | Intron | NP_114427.1 | ||||
XM_006721096.3 | Intron | XP_006721159.1 | ||||
XM_017023757.1 | Intron | XP_016879246.1 | ||||
XM_017023758.1 | Intron | XP_016879247.1 | ||||
XM_017023759.1 | Intron | XP_016879248.1 |