Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCAGGGAGGACTCGAGCATGGTGC[A/C]TGGGCATCAGGACCAAACGGATGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613141 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DTX2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DTX2 - deltex E3 ubiquitin ligase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102594.1 | Intron | NP_001096064.1 | ||||
NM_001102595.1 | Intron | NP_001096065.1 | ||||
NM_001102596.1 | Intron | NP_001096066.1 | ||||
NM_020892.2 | Intron | NP_065943.2 | ||||
XM_005250131.1 | Intron | XP_005250188.1 | ||||
XM_005250132.1 | Intron | XP_005250189.1 | ||||
XM_011515772.1 | Intron | XP_011514074.1 | ||||
XM_011515773.2 | Intron | XP_011514075.1 | ||||
XM_011515774.1 | Intron | XP_011514076.1 | ||||
XM_011515775.1 | Intron | XP_011514077.1 | ||||
XM_017011723.1 | Intron | XP_016867212.1 | ||||
XM_017011724.1 | Intron | XP_016867213.1 | ||||
XM_017011725.1 | Intron | XP_016867214.1 | ||||
XM_017011726.1 | Intron | XP_016867215.1 | ||||
XM_017011727.1 | Intron | XP_016867216.1 | ||||
XM_017011728.1 | Intron | XP_016867217.1 | ||||
XM_017011729.1 | Intron | XP_016867218.1 | ||||
XM_017011730.1 | Intron | XP_016867219.1 |
LOC105375355 - uroplakin-3b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |