Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCCAGTGCAACCGCTTGTTATA[A/G]CACAACCCTTCTAGTTCTAACCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603626 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PCDH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PCDH1 - protocadherin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278613.1 | Intron | NP_001265542.1 | ||||
NM_001278615.1 | Intron | NP_001265544.1 | ||||
NM_002587.4 | Intron | NP_002578.2 | ||||
NM_032420.3 | Intron | NP_115796.2 | ||||
XM_005268452.2 | Intron | XP_005268509.1 | ||||
XM_005268454.4 | Intron | XP_005268511.1 | ||||
XM_005268455.2 | Intron | XP_005268512.1 | ||||
XM_017009517.1 | Intron | XP_016865006.1 | ||||
XM_017009518.1 | Intron | XP_016865007.1 |
Set Membership: |
HapMap |