Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGTTCTTCTCAGTCTTCAAGGC[A/G]CATTGCCAATATCCTCTCATTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616426 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP192 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CEP192 - centrosomal protein 192 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032142.3 | Intron | NP_115518.3 | ||||
XM_005258107.3 | Intron | XP_005258164.1 | ||||
XM_005258109.3 | Intron | XP_005258166.1 | ||||
XM_005258110.1 | Intron | XP_005258167.1 | ||||
XM_006722326.3 | Intron | XP_006722389.1 | ||||
XM_006722327.3 | Intron | XP_006722390.1 | ||||
XM_006722330.3 | Intron | XP_006722393.1 | ||||
XM_011525673.2 | Intron | XP_011523975.1 | ||||
XM_011525675.2 | Intron | XP_011523977.1 | ||||
XM_017025803.1 | Intron | XP_016881292.1 | ||||
XM_017025804.1 | Intron | XP_016881293.1 | ||||
XM_017025805.1 | Intron | XP_016881294.1 | ||||
XM_017025806.1 | Intron | XP_016881295.1 | ||||
XM_017025807.1 | Intron | XP_016881296.1 |