Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602396 | ||||||||||||||||||||
Literature Links: |
ANXA8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANXA8 - annexin A8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040084.2 | Intron | NP_001035173.1 | ||||
NM_001271702.1 | Intron | NP_001258631.1 | ||||
NM_001271703.1 | Intron | NP_001258632.1 | ||||
XM_006717951.3 | Intron | XP_006718014.1 | ||||
XM_011540096.1 | Intron | XP_011538398.1 | ||||
XM_011540098.1 | Intron | XP_011538400.1 | ||||
XM_011540099.1 | Intron | XP_011538401.1 | ||||
XM_011540101.2 | Intron | XP_011538403.1 |
BMS1P6 - BMS1, ribosome biogenesis factor pseudogene 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |