Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614124 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPCPD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GPCPD1 - glycerophosphocholine phosphodiesterase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_019593.3 | 3923 | UTR 3 | NP_062539.1 | |||
XM_005260758.4 | 3923 | UTR 3 | XP_005260815.1 | |||
XM_005260761.1 | 3923 | UTR 3 | XP_005260818.1 | |||
XM_006723596.3 | 3923 | UTR 3 | XP_006723659.1 |
Set Membership: |
HapMap |