Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613781 MIM: 601955 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC125 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC125 - coiled-coil domain containing 125 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297696.1 | Intron | NP_001284625.1 | ||||
NM_001297697.1 | Intron | NP_001284626.1 | ||||
NM_176816.4 | Intron | NP_789786.2 | ||||
XM_005248458.4 | Intron | XP_005248515.1 | ||||
XM_005248459.4 | Intron | XP_005248516.1 | ||||
XM_005248461.3 | Intron | XP_005248518.1 | ||||
XM_005248463.3 | Intron | XP_005248520.1 | ||||
XM_005248464.3 | Intron | XP_005248521.1 | ||||
XM_006714569.3 | Intron | XP_006714632.1 | ||||
XM_006714570.3 | Intron | XP_006714633.1 | ||||
XM_011543255.2 | Intron | XP_011541557.1 | ||||
XM_011543256.2 | Intron | XP_011541558.1 | ||||
XM_011543257.2 | Intron | XP_011541559.1 | ||||
XM_011543258.2 | Intron | XP_011541560.1 | ||||
XM_011543259.2 | Intron | XP_011541561.1 | ||||
XM_011543260.2 | Intron | XP_011541562.1 | ||||
XM_017009206.1 | Intron | XP_016864695.1 | ||||
XM_017009207.1 | Intron | XP_016864696.1 | ||||
XM_017009208.1 | Intron | XP_016864697.1 | ||||
XM_017009209.1 | Intron | XP_016864698.1 | ||||
XM_017009210.1 | Intron | XP_016864699.1 | ||||
XM_017009211.1 | Intron | XP_016864700.1 |
CDK7 - cyclin dependent kinase 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001324069.1 | Intron | NP_001310998.1 | ||||
NM_001324070.1 | Intron | NP_001310999.1 | ||||
NM_001324071.1 | Intron | NP_001311000.1 | ||||
NM_001324072.1 | Intron | NP_001311001.1 | ||||
NM_001324074.1 | Intron | NP_001311003.1 | ||||
NM_001324075.1 | Intron | NP_001311004.1 | ||||
NM_001324077.1 | Intron | NP_001311006.1 | ||||
NM_001324078.1 | Intron | NP_001311007.1 | ||||
NM_001799.3 | Intron | NP_001790.1 | ||||
XM_011543094.2 | Intron | XP_011541396.1 |
Set Membership: |
HapMap |