Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM39B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TMEM39B - transmembrane protein 39B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319677.1 | Intron | NP_001306606.1 | ||||
NM_001319678.1 | Intron | NP_001306607.1 | ||||
NM_001319679.1 | Intron | NP_001306608.1 | ||||
NM_018056.3 | Intron | NP_060526.2 | ||||
XM_005270988.2 | Intron | XP_005271045.1 | ||||
XM_006710724.1 | Intron | XP_006710787.1 | ||||
XM_006710725.3 | Intron | XP_006710788.1 | ||||
XM_011541681.2 | Intron | XP_011539983.1 | ||||
XM_011541683.1 | Intron | XP_011539985.1 | ||||
XM_011541684.2 | Intron | XP_011539986.1 | ||||
XM_017001595.1 | Intron | XP_016857084.1 |