Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610982 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
INF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
INF2 - inverted formin, FH2 and WH2 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031714.3 | Intron | NP_001026884.3 | ||||
NM_022489.3 | Intron | NP_071934.3 | ||||
NM_032714.2 | Intron | NP_116103.1 | ||||
XM_005268004.4 | Intron | XP_005268061.1 | ||||
XM_005268005.4 | Intron | XP_005268062.1 | ||||
XM_017021595.1 | Intron | XP_016877084.1 |