Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616446 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LEXM PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LEXM - lymphocyte expansion molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110533.1 | Intron | NP_001104003.1 | ||||
NM_152607.2 | Intron | NP_689820.2 | ||||
XM_017000480.1 | Intron | XP_016855969.1 | ||||
XM_017000481.1 | Intron | XP_016855970.1 |
TTC22 - tetratricopeptide repeat domain 22 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |