Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACACAATACAAATAATGAACCCAA[A/G]AACTAAATGAGAAAATGTTCATGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605070 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EEA1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EEA1 - early endosome antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003566.3 | 7750 | UTR 3 | NP_003557.2 | |||
XM_011538814.2 | 7750 | UTR 3 | XP_011537116.1 | |||
XM_011538815.2 | 7750 | UTR 3 | XP_011537117.1 | |||
XM_017020018.1 | 7750 | UTR 3 | XP_016875507.1 |
PLEKHG7 - pleckstrin homology and RhoGEF domain containing G7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004330.2 | 7750 | Intron | NP_001004330.1 |
Set Membership: |
HapMap |