Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614804 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DMRTA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DMRTA2 - DMRT like family A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032110.2 | 2389 | Silent Mutation | CTA,CTG | L,L 420 | NP_115486.1 | |
XM_011541937.2 | 2389 | Silent Mutation | CTA,CTG | L,L 420 | XP_011540239.1 |