Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
36 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606649 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIVEP3 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HIVEP3 - human immunodeficiency virus type I enhancer binding protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127714.2 | 12157 | UTR 3 | NP_001121186.1 | |||
NM_024503.4 | 12157 | UTR 3 | NP_078779.2 | |||
XM_011541884.2 | 12157 | Intron | XP_011540186.1 | |||
XM_011541885.2 | 12157 | Intron | XP_011540187.1 | |||
XM_011541886.2 | 12157 | Intron | XP_011540188.1 | |||
XM_011541887.2 | 12157 | Intron | XP_011540189.1 | |||
XM_017001992.1 | 12157 | Intron | XP_016857481.1 | |||
XM_017001993.1 | 12157 | Intron | XP_016857482.1 | |||
XM_017001994.1 | 12157 | Intron | XP_016857483.1 |