Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605449 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GSTA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Japanese
|
CHB (Han Chinese) - Not Available | ||||||
AFR
|
Chinese
|
JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GSTA3 - glutathione S-transferase alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000847.4 | 124 | Missense Mutation | ATT,CTT | I,L 71 | NP_000838.3 | |
XM_006715072.3 | 124 | Missense Mutation | ATT,CTT | I,L 21 | XP_006715135.1 | |
XM_011514533.2 | 124 | Intron | XP_011512835.1 | |||
XM_017010794.1 | 124 | Missense Mutation | ATT,CTT | I,L 21 | XP_016866283.1 |
Set Membership: |
DME Validated Inventoried |