Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180386 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LMO3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LMO3 - LIM domain only 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001395.2 | Intron | NP_001001395.1 | ||||
NM_001243609.1 | Intron | NP_001230538.1 | ||||
NM_001243610.1 | Intron | NP_001230539.1 | ||||
NM_001243611.1 | Intron | NP_001230540.1 | ||||
NM_001243612.1 | Intron | NP_001230541.1 | ||||
NM_001243613.1 | Intron | NP_001230542.1 | ||||
NM_018640.4 | Intron | NP_061110.2 | ||||
XM_006719110.3 | Intron | XP_006719173.1 | ||||
XM_006719111.2 | Intron | XP_006719174.1 | ||||
XM_011520762.1 | Intron | XP_011519064.1 | ||||
XM_011520763.1 | Intron | XP_011519065.1 | ||||
XM_017019643.1 | Intron | XP_016875132.1 |