Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTCTGGAAGCAGAAGAGTAGGAG[G/T]GTTGGTGGGTGAAATTCTGAGATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180902 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RYR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RYR2 - ryanodine receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001035.2 | Intron | NP_001026.2 | ||||
XM_006711802.3 | Intron | XP_006711865.1 | ||||
XM_006711803.3 | Intron | XP_006711866.1 | ||||
XM_006711804.3 | Intron | XP_006711867.1 | ||||
XM_006711805.3 | Intron | XP_006711868.1 | ||||
XM_006711806.3 | Intron | XP_006711869.1 | ||||
XM_006711807.3 | Intron | XP_006711870.1 | ||||
XM_006711808.3 | Intron | XP_006711871.1 | ||||
XM_006711809.3 | Intron | XP_006711872.1 | ||||
XM_006711810.3 | Intron | XP_006711873.1 | ||||
XM_017002028.1 | Intron | XP_016857517.1 |
Set Membership: |
HapMap |