Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602760 MIM: 602764 MIM: 604540 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KRT32 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KRT32 - keratin 32 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KRT35 - keratin 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002280.4 | 2973 | Missense Mutation | CCC,GCC | P,A 443 | NP_002271.3 | |
XM_011524794.1 | 2973 | Missense Mutation | CCC,GCC | P,A 443 | XP_011523096.1 |
KRT36 - keratin 36 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |