Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTATAATTTATTAAGCAAACAA[A/G]GGATCAAATGTCTCTCCATCCACCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 159552 | ||||||||||||||||||||
Literature Links: |
ADAMTSL4-AS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAMTSL4-AS1 - ADAMTSL4 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCL1 - BCL2 family apoptosis regulator | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001197320.1 | 2289 | UTR 3 | NP_001184249.1 | |||
NM_021960.4 | 2289 | UTR 3 | NP_068779.1 | |||
NM_182763.2 | 2289 | UTR 3 | NP_877495.1 |