Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAGATATACATTTACACAAAGAA[A/G]TGGGTATTGTGAAAGAAAACATATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105370888 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105370888 - uncharacterized LOC105370888 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM202 - transmembrane protein 202 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080462.1 | Intron | NP_001073931.1 | ||||
XM_011521497.1 | Intron | XP_011519799.1 | ||||
XM_011521499.1 | Intron | XP_011519801.1 |
Set Membership: |
HapMap |