Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 104311 | ||||||||||||||||||||
Literature Links: |
PSEN1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PSEN1 - presenilin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000021.3 | Intron | NP_000012.1 | ||||
NM_007318.2 | Intron | NP_015557.2 | ||||
XM_005267864.2 | Intron | XP_005267921.1 | ||||
XM_005267866.1 | Intron | XP_005267923.1 | ||||
XM_011536971.2 | Intron | XP_011535273.1 | ||||
XM_011536972.2 | Intron | XP_011535274.1 | ||||
XM_011536973.1 | Intron | XP_011535275.1 | ||||
XM_011536974.1 | Intron | XP_011535276.1 |