Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTAGCCGCTTGCGTGTTGAAAC[C/T]TCTTCCCTCTATGTTCGCCGGACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
34 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613455 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIA3 - MIA family member 3, ER export factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300867.1 | Intron | NP_001287796.1 | ||||
NM_001324062.1 | Intron | NP_001310991.1 | ||||
NM_001324063.1 | Intron | NP_001310992.1 | ||||
NM_001324064.1 | Intron | NP_001310993.1 | ||||
NM_001324065.1 | Intron | NP_001310994.1 | ||||
NM_198551.3 | Intron | NP_940953.2 | ||||
XM_006711304.3 | Intron | XP_006711367.2 | ||||
XM_011509513.2 | Intron | XP_011507815.2 | ||||
XM_017001243.1 | Intron | XP_016856732.1 |