Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
A1CF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
A1CF - APOBEC1 complementation factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198818.1 | 3586 | UTR 3 | NP_001185747.1 | |||
NM_001198819.1 | 3586 | UTR 3 | NP_001185748.1 | |||
NM_001198820.1 | 3586 | UTR 3 | NP_001185749.1 | |||
NM_014576.3 | 3586 | UTR 3 | NP_055391.2 | |||
NM_138932.2 | 3586 | UTR 3 | NP_620310.1 | |||
NM_138933.2 | 3586 | UTR 3 | NP_620311.1 |