Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
43 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf100 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf100 - chromosome 1 open reading frame 100 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012970.2 | 758 | Intron | NP_001012988.1 | |||
NM_001276348.1 | 758 | Intron | NP_001263277.1 | |||
NM_001276349.1 | 758 | Intron | NP_001263278.1 | |||
XM_011544124.2 | 758 | Intron | XP_011542426.1 | |||
XM_011544125.2 | 758 | Intron | XP_011542427.1 | |||
XM_011544128.2 | 758 | UTR 5 | XP_011542430.1 | |||
XM_011544129.2 | 758 | Intron | XP_011542431.1 | |||
XM_011544130.2 | 758 | Intron | XP_011542432.1 | |||
XM_017000571.1 | 758 | Intron | XP_016856060.1 | |||
XM_017000572.1 | 758 | UTR 5 | XP_016856061.1 |