Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601574 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HEATR9 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HEATR9 - HEAT repeat containing 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321395.1 | 1531 | Missense Mutation | TCT,TTT | S,F 440 | NP_001308324.1 | |
NM_152781.3 | 1531 | Missense Mutation | TCT,TTT | S,F 480 | NP_689994.2 | |
XM_005278233.2 | 1531 | Missense Mutation | TCT,TTT | S,F 446 | XP_005278290.1 | |
XM_005278235.1 | 1531 | Missense Mutation | TCT,TTT | S,F 406 | XP_005278292.1 | |
XM_006721802.3 | 1531 | Missense Mutation | TCT,TTT | S,F 298 | XP_006721865.1 | |
XM_011524586.2 | 1531 | Missense Mutation | TCT,TTT | S,F 446 | XP_011522888.1 | |
XM_017024424.1 | 1531 | UTR 3 | XP_016879913.1 |
TAF15 - TATA-box binding protein associated factor 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |