Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607806 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OTOP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OTOP1 - otopetrin 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM128 - transmembrane protein 128 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297551.1 | 1313 | Intron | NP_001284480.1 | |||
NM_001297552.1 | 1313 | Intron | NP_001284481.1 | |||
NM_032927.3 | 1313 | Intron | NP_116316.1 | |||
XM_005248034.2 | 1313 | UTR 3 | XP_005248091.1 |