Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGATTACCTGTGATTAAACATAA[A/G]TCACACAATAAAATAGGATCTCCTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612197 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABHD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ABHD3 - abhydrolase domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308256.1 | Intron | NP_001295185.1 | ||||
NM_001308257.1 | Intron | NP_001295186.1 | ||||
NM_138340.4 | Intron | NP_612213.2 | ||||
XM_011525847.2 | Intron | XP_011524149.1 | ||||
XM_017025571.1 | Intron | XP_016881060.1 | ||||
XM_017025572.1 | Intron | XP_016881061.1 | ||||
XM_017025573.1 | Intron | XP_016881062.1 | ||||
XM_017025574.1 | Intron | XP_016881063.1 |
Set Membership: |
HapMap |