Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGTTGAGTAACTTTCCCAAGATC[A/C]TACTGGTAATAAGTGATGATGCCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
33 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601924 MIM: 605254 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COPA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
COPA - coatomer protein complex subunit alpha | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCSTN - nicastrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001290184.1 | Intron | NP_001277113.1 | ||||
NM_001290186.1 | Intron | NP_001277115.1 | ||||
NM_015331.2 | Intron | NP_056146.1 | ||||
XM_005245053.4 | Intron | XP_005245110.1 | ||||
XM_011509363.2 | Intron | XP_011507665.1 |