Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603868 MIM: 613262 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB27A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB27A - RAB27A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004580.4 | 2716 | UTR 3 | NP_004571.2 | |||
NM_183234.2 | 2716 | UTR 3 | NP_899057.1 | |||
NM_183235.2 | 2716 | UTR 3 | NP_899058.1 | |||
NM_183236.2 | 2716 | UTR 3 | NP_899059.1 | |||
XM_005254576.4 | 2716 | UTR 3 | XP_005254633.1 | |||
XM_011521852.1 | 2716 | UTR 3 | XP_011520154.1 | |||
XM_011521854.1 | 2716 | UTR 3 | XP_011520156.1 | |||
XM_011521855.2 | 2716 | UTR 3 | XP_011520157.1 | |||
XM_011521856.2 | 2716 | UTR 3 | XP_011520158.1 |
RSL24D1 - ribosomal L24 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |