Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCTAGTGTTAATTTAGAAATATT[A/G]CACCGTGGCTGGGCACTGTGGCTCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612061 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105375218 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC105375218 - uncharacterized LOC105375218 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR550A1 - microRNA 550a-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR550B1 - microRNA 550b-1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNRF2 - zinc and ring finger 2, E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_147128.3 | Intron | NP_667339.1 |
Set Membership: |
HapMap |