Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTCCTCTAGGTCCTTTAGGAGG[C/T]CATAGACGTTGCTGTCAGAGGCGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603515 MIM: 139250 | ||||||||||||||||||||
Literature Links: |
CSHL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CSHL1 - chorionic somatomammotropin hormone like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GH1 - growth hormone 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000515.4 | 489 | Missense Mutation | GAC,GGC | D,G 138 | NP_000506.2 | |
NM_022559.3 | 489 | Missense Mutation | GAC,GGC | D,G 123 | NP_072053.1 | |
NM_022560.3 | 489 | Missense Mutation | GAC,GGC | D,G 98 | NP_072054.1 |