Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGTTAAATATGTGAGTTAAAAAA[A/G]ATTTTAAACCATAACCTATGAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609544 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C16orf62 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C16orf62 - chromosome 16 open reading frame 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300743.1 | Intron | NP_001287672.1 | ||||
NM_020314.5 | Intron | NP_064710.4 | ||||
XM_005255435.3 | Intron | XP_005255492.1 | ||||
XM_006721065.3 | Intron | XP_006721128.1 | ||||
XM_011545896.2 | Intron | XP_011544198.1 | ||||
XM_011545897.2 | Intron | XP_011544199.1 |
CCP110 - centriolar coiled-coil protein 110 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |