Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAGTAAATGGGGAGATCTCAGATT[A/C]TTTTTTTTGAGACAGAGTCTCGCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614630 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADGB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ADGB - androglobin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024694.3 | Intron | NP_078970.3 | ||||
XM_006715566.3 | Intron | XP_006715629.1 | ||||
XM_011536125.2 | Intron | XP_011534427.1 | ||||
XM_011536126.1 | Intron | XP_011534428.1 | ||||
XM_011536127.2 | Intron | XP_011534429.1 | ||||
XM_011536128.2 | Intron | XP_011534430.1 | ||||
XM_011536130.2 | Intron | XP_011534432.1 | ||||
XM_011536131.2 | Intron | XP_011534433.1 | ||||
XM_011536132.2 | Intron | XP_011534434.1 | ||||
XM_011536134.2 | Intron | XP_011534436.1 | ||||
XM_011536135.2 | Intron | XP_011534437.1 | ||||
XM_017011314.1 | Intron | XP_016866803.1 | ||||
XM_017011315.1 | Intron | XP_016866804.1 |