Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTCAGAGAGGCCAGGATCATCTCC[C/T]CGTGTCACGACCCTTCATCTCATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601624 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
FCN2 - ficolin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004108.2 | Intron | NP_004099.2 | ||||
NM_015837.2 | Intron | NP_056652.1 | ||||
XM_006717015.3 | Intron | XP_006717078.1 | ||||
XM_011518392.2 | Intron | XP_011516694.1 |
LOC101448202 - uncharacterized LOC101448202 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |