Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606682 MIM: 602254 MIM: 612747 | ||||||||||||||||||||
Literature Links: |
HPS4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HPS4 - HPS4, biogenesis of lysosomal organelles complex 3 subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRRD - SRR1 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013694.2 | 1543 | Intron | NP_001013716.2 | |||
XM_011530178.2 | 1543 | UTR 3 | XP_011528480.1 | |||
XM_017028799.1 | 1543 | Intron | XP_016884288.1 |
TFIP11 - tuftelin interacting protein 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008697.1 | 1543 | Missense Mutation | CCC,CTC | P,L 801 | NP_001008697.1 | |
NM_012143.2 | 1543 | Missense Mutation | CCC,CTC | P,L 801 | NP_036275.1 | |
XM_011530079.1 | 1543 | Missense Mutation | CCC,CTC | P,L 801 | XP_011528381.1 | |
XM_017028735.1 | 1543 | Missense Mutation | CCC,CTC | P,L 801 | XP_016884224.1 |