Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614570 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KIF18B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KIF18B - kinesin family member 18B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001264573.1 | 2692 | UTR 3 | NP_001251503.1 | |||
NM_001265577.1 | 2692 | Silent Mutation | GCA,GCG | A,A 841 | NP_001252506.1 | |
XM_011524385.2 | 2692 | Silent Mutation | GCA,GCG | A,A 862 | XP_011522687.1 | |
XM_011524386.2 | 2692 | Silent Mutation | GCA,GCG | A,A 853 | XP_011522688.1 | |
XM_011524387.2 | 2692 | Silent Mutation | GCA,GCG | A,A 853 | XP_011522689.1 | |
XM_011524388.2 | 2692 | Silent Mutation | GCA,GCG | A,A 850 | XP_011522690.1 | |
XM_011524389.2 | 2692 | UTR 3 | XP_011522691.1 | |||
XM_011524390.2 | 2692 | Silent Mutation | GCA,GCG | A,A 827 | XP_011522692.1 | |
XM_011524391.2 | 2692 | Intron | XP_011522693.1 |
MIR6783 - microRNA 6783 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |