Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
62 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 188360 MIM: 188340 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD1B PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CD1B - CD1b molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001764.2 | 501 | Intron | NP_001755.1 | |||
XM_011510119.2 | 501 | Intron | XP_011508421.1 | |||
XM_017002784.1 | 501 | Intron | XP_016858273.1 | |||
XM_017002785.1 | 501 | Intron | XP_016858274.1 | |||
XM_017002786.1 | 501 | Intron | XP_016858275.1 |
CD1C - CD1c molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001765.2 | 501 | Missense Mutation | AAC,ACC | N,T 70 | NP_001756.2 | |
XM_005245579.4 | 501 | Missense Mutation | AAC,ACC | N,T 70 | XP_005245636.1 |
Set Membership: |
HapMap |