Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 163730 | ||||||||||||||||||||
Literature Links: |
NOS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NOS2 - nitric oxide synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000625.4 | 3389 | UTR 3 | NP_000616.3 | |||
XM_011524859.2 | 3389 | UTR 3 | XP_011523161.1 | |||
XM_011524860.2 | 3389 | UTR 3 | XP_011523162.1 | |||
XM_011524861.1 | 3389 | UTR 3 | XP_011523163.1 | |||
XM_011524862.1 | 3389 | UTR 3 | XP_011523164.1 |