Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606781 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TKT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TKT - transketolase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001064.3 | 2029 | Intron | NP_001055.1 | |||
NM_001135055.2 | 2029 | UTR 3 | NP_001128527.1 | |||
NM_001258028.1 | 2029 | Intron | NP_001244957.1 | |||
XM_011534054.1 | 2029 | UTR 3 | XP_011532356.1 | |||
XM_011534055.1 | 2029 | Intron | XP_011532357.1 |
Set Membership: |
HapMap |