Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC45A4 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SLC45A4 - solute carrier family 45 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080431.2 | 3084 | UTR 3 | NP_001073900.1 | |||
NM_001286646.1 | 3084 | UTR 3 | NP_001273575.1 | |||
NM_001286648.1 | 3084 | Intron | NP_001273577.1 | |||
XM_011517169.2 | 3084 | UTR 3 | XP_011515471.1 | |||
XM_011517170.2 | 3084 | UTR 3 | XP_011515472.1 | |||
XM_011517171.2 | 3084 | UTR 3 | XP_011515473.1 | |||
XM_011517172.2 | 3084 | UTR 3 | XP_011515474.1 | |||
XM_011517173.2 | 3084 | Intron | XP_011515475.1 | |||
XM_011517174.2 | 3084 | UTR 3 | XP_011515476.1 | |||
XM_011517176.2 | 3084 | UTR 3 | XP_011515478.1 | |||
XM_011517177.2 | 3084 | UTR 3 | XP_011515479.1 | |||
XM_011517178.2 | 3084 | UTR 3 | XP_011515480.1 | |||
XM_011517179.2 | 3084 | UTR 3 | XP_011515481.1 | |||
XM_017013648.1 | 3084 | UTR 3 | XP_016869137.1 | |||
XM_017013649.1 | 3084 | UTR 3 | XP_016869138.1 | |||
XM_017013650.1 | 3084 | UTR 3 | XP_016869139.1 |
Set Membership: |
HapMap |