Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107580 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TFAP2A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
TFAP2A - transcription factor AP-2 alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032280.2 | 1485 | Silent Mutation | AAC,AAT | N,N 413 | NP_001027451.1 | |
NM_001042425.1 | 1485 | Silent Mutation | AAC,AAT | N,N 415 | NP_001035890.1 | |
NM_003220.2 | 1485 | Silent Mutation | AAC,AAT | N,N 419 | NP_003211.1 | |
XM_006715175.2 | 1485 | Silent Mutation | AAC,AAT | N,N 464 | XP_006715238.1 | |
XM_011514833.2 | 1485 | Silent Mutation | AAC,AAT | N,N 369 | XP_011513135.1 | |
XM_017011232.1 | 1485 | Silent Mutation | AAC,AAT | N,N 501 | XP_016866721.1 |
Set Membership: |
HapMap |